MAN182106773 Manchester Organics

2-(Methylthio)thiophene

  • MDL number MFCD00052382
  • Molecular Weight : 130.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/V17023
Availability details for
Quantity
  • Availability
  • Add to Cart