MAN182106909 Manchester Organics

3,4-Dihydroxy-5-methoxybenzoic acid

  • MDL number MFCD00016518
  • Molecular Weight : 184.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V17733

Safety Information

MSDS https://www.manchesterorganics.com/V17733
Availability details for
Quantity
  • Availability
  • Add to Cart