MAN182106932 Manchester Organics

Imidazo[1,2-b]pyridazin-3-ylamine

  • MDL number MFCD09832520
  • Molecular Weight : 134.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C17859

Safety Information

MSDS https://www.manchesterorganics.com/C17859
Availability details for
Quantity
  • Availability
  • Add to Cart