MAN182106947 Manchester Organics

2,4,6-Trichloro-3-nitropyridine

  • MDL number MFCD09264535
  • Molecular Weight : 227.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z17194

Safety Information

MSDS https://www.manchesterorganics.com/Z17194
Availability details for
Quantity
  • Availability
  • Add to Cart