MAN182106961 Manchester Organics

3-Bromopyridine N-oxide

  • MDL number MFCD01646056
  • Cas Number : 2402-97-3
  • Molecular Weight : 174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N16378
Availability details for
Quantity
  • Availability
  • Add to Cart