MAN182106971 Manchester Organics

Ethyl dichlorophosphate

  • MDL number MFCD00002069
  • Cas Number : 1498-51-7
  • Molecular Weight : 162.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/V17449
Availability details for
Quantity
  • Availability
  • Add to Cart