MAN182107037 Manchester Organics

2-Acetyl-5-nitrobenzo[b]furan

  • MDL number MFCD00051634
  • Cas Number : 23136-39-2
  • Molecular Weight : 205.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/J10197
Availability details for
Quantity
  • Availability
  • Add to Cart