MAN182107046 Manchester Organics

3,4-Dichlorobenzenesulfonamide

  • MDL number MFCD01314052
  • Cas Number : 23815-28-3
  • Molecular Weight : 226.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/P15007
Availability details for
Quantity
  • Availability
  • Add to Cart