MAN182107091 Manchester Organics

3-Fluoro-4-methoxybenzenesulfonamide

  • MDL number MFCD02091381
  • Cas Number : 874767-60-9
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/U10870
Availability details for
Quantity
  • Availability
  • Add to Cart