MAN182107105 Manchester Organics

2-Methyl-4,4,4-trifluorobutyric acid

  • MDL number MFCD00190640
  • Molecular Weight : 156.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R16925

Safety Information

MSDS https://www.manchesterorganics.com/R16925
Availability details for
Quantity
  • Availability
  • Add to Cart