MAN182107142 Manchester Organics

2-Butylthiophene

  • MDL number MFCD00022534
  • Molecular Weight : 140.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y17761

Safety Information

MSDS https://www.manchesterorganics.com/Y17761
Availability details for
Quantity
  • Availability
  • Add to Cart