MAN182107164 Manchester Organics

4-Bromo-2-(trifluoromethoxy)phenol

  • MDL number MFCD09037798
  • Cas Number : 690264-39-2
  • Molecular Weight : 257.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2-(trifluoromethoxy)phenol
Inchi Code 1S/C7H4BrF3O2/c8-4-1-2-5(12)6(3-4)13-7(9,10)11/h1-3,12H
Inchi Key GYXNTPVMXGBRQW-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin GB
Product Link https://www.manchesterorganics.com/D16559
Availability details for
Quantity
  • Availability
  • Add to Cart