MAN182107169 Manchester Organics

3-Bromoimidazo[1,2-b]pyridazine

  • MDL number MFCD09757672
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J17771
Availability details for
Quantity
  • Availability
  • Add to Cart