MAN182107178 Manchester Organics

1-(5-Phenyl-1,3,4-oxadiazol-2-yl)methylamine

  • MDL number MFCD06490519
  • Molecular Weight : 175.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U17354

Safety Information

MSDS https://www.manchesterorganics.com/U17354
Availability details for
Quantity
  • Availability
  • Add to Cart