MAN182107190 Manchester Organics

7-(Trifluoromethyl)quinoline

  • MDL number MFCD00833760
  • Cas Number : 325-14-4
  • Molecular Weight : 197.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/R10771
Availability details for
Quantity
  • Availability
  • Add to Cart