MAN182107195 Manchester Organics

2,4,6-Trifluoropyrimidine

  • MDL number MFCD00039705
  • Cas Number : 696-82-2
  • Molecular Weight : 134.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/X17831
Availability details for
Quantity
  • Availability
  • Add to Cart