MAN182107208 Manchester Organics

4-Amino-6-hydroxypyrimidine

  • MDL number MFCD00051502
  • Molecular Weight : 111.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C16700

Safety Information

MSDS https://www.manchesterorganics.com/C16700
Availability details for
Quantity
  • Availability
  • Add to Cart