MAN182107212 Manchester Organics

2-Methoxy-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD01862091
  • Molecular Weight : 220.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B17881

Safety Information

MSDS https://www.manchesterorganics.com/B17881
Availability details for
Quantity
  • Availability
  • Add to Cart