MAN182107213 Manchester Organics

2,3-Difluoro-5-(trifluoromethyl)pyridine

  • MDL number MFCD04972837
  • Cas Number : 89402-42-6
  • Molecular Weight : 183.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/C16368
Availability details for
Quantity
  • Availability
  • Add to Cart