MAN182107227 Manchester Organics

4-Chloro-2-(trifluoromethyl)benzoic acid

  • MDL number MFCD03094162
  • Cas Number : 142994-09-0
  • Molecular Weight : 224.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/T17802
Availability details for
Quantity
  • Availability
  • Add to Cart