MAN182107279 Manchester Organics

2,3-Difluoro-4-methoxybenzaldehyde

  • MDL number MFCD04115981
  • Cas Number : 256417-11-5
  • Molecular Weight : 172.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/J17865
Availability details for
Quantity
  • Availability
  • Add to Cart