MAN182107290 Manchester Organics

4-Bromo-2-nitrophenylhydrazine hydrochloride

  • MDL number MFCD09260482
  • Molecular Weight : 268.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B10735

Safety Information

MSDS https://www.manchesterorganics.com/B10735
Availability details for
Quantity
  • Availability
  • Add to Cart