MAN182107321 Manchester Organics

1-[2-(Trifluoromethyl)phenyl]ethan-1-ol

  • MDL number MFCD00004513
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L17963

Safety Information

MSDS https://www.manchesterorganics.com/L17963
Availability details for
Quantity
  • Availability
  • Add to Cart