MAN182107336 Manchester Organics

2,3,4,5-Tetrahydro-benzo[b][1,4]diazepine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD09832499
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C18001

Safety Information

MSDS https://www.manchesterorganics.com/C18001
Availability details for
Quantity
  • Availability
  • Add to Cart