MAN182107372 Manchester Organics

7-Fluoro-2,3,4,9-tetrahydro-1H-beta-carboline

  • MDL number MFCD09264527
  • Molecular Weight : 190.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B10829

Safety Information

MSDS https://www.manchesterorganics.com/B10829
Availability details for
Quantity
  • Availability
  • Add to Cart