MAN182107396 Manchester Organics

5-(4-Methoxy-phenyl)-3,4-dihydro-2H-pyrrole

  • MDL number MFCD09749694
  • Molecular Weight : 175.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J18007

Safety Information

MSDS https://www.manchesterorganics.com/J18007
Availability details for
Quantity
  • Availability
  • Add to Cart