MAN182107412 Manchester Organics

2,4-Difluoro-3-methylbenzyl alcohol

  • MDL number MFCD07777159
  • Cas Number : 847502-91-4
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/W17947
Availability details for
Quantity
  • Availability
  • Add to Cart