MAN182107426 Manchester Organics

2-Hydroxy-3-aminopyridine

  • MDL number MFCD03411556
  • Molecular Weight : 110.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z16530

Safety Information

MSDS https://www.manchesterorganics.com/Z16530
Availability details for
Quantity
  • Availability
  • Add to Cart