MAN182107434 Manchester Organics

2,6-Dimethyl-4-aminoanisole

  • MDL number MFCD01907075
  • Cas Number : 39785-37-0
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/B17954
Availability details for
Quantity
  • Availability
  • Add to Cart