MAN182107437 Manchester Organics

4,5-Difluoro-2-methoxybenzonitrile

  • MDL number MFCD06660346
  • Molecular Weight : 169.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A16604
Availability details for
Quantity
  • Availability
  • Add to Cart