MAN182107471 Manchester Organics

3-Amino-4-(methylthio)benzotrifluoride

  • MDL number MFCD00052715
  • Cas Number : 207974-07-0
  • Molecular Weight : 207.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(methylsulfanyl)-5-(trifluoromethyl)aniline
Inchi Code 1S/C8H8F3NS/c1-13-7-3-2-5(4-6(7)12)8(9,10)11/h2-4H,12H2,1H3
Inchi Key OQFOLJGOVFYDHZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/H10008
Availability details for
Quantity
  • Availability
  • Add to Cart