MAN182107482 Manchester Organics

3-Nitro-4-ethoxypyridine

  • MDL number MFCD00234959
  • Cas Number : 1796-84-5
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-ethoxy-3-nitropyridine
Inchi Code 1S/C7H8N2O3/c1-2-12-7-3-4-8-5-6(7)9(10)11/h3-5H,2H2,1H3
Inchi Key ABOSMHRLVUWEMT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/E16040
Availability details for
Quantity
  • Availability
  • Add to Cart