MAN182107493 Manchester Organics

2,3-Difluoro-4-methoxybenzoic acid

  • MDL number MFCD03428569
  • Molecular Weight : 188.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J16186

Safety Information

MSDS https://www.manchesterorganics.com/J16186
Availability details for
Quantity
  • Availability
  • Add to Cart