MAN182107587 Manchester Organics

3-Methylsulfonylbenzoic acid

  • MDL number MFCD00216488
  • Cas Number : 5345-27-7
  • Molecular Weight : 200.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/L17986
Availability details for
Quantity
  • Availability
  • Add to Cart