MAN182107610 Manchester Organics

N-Benzyloxycarbonylglycine thioamide

  • MDL number MFCD03452777
  • Cas Number : 49548-40-5
  • Molecular Weight : 224.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y10210
Availability details for
Quantity
  • Availability
  • Add to Cart