MAN182107611 Manchester Organics

3,5-Bis(trifluoromethyl)benzhydrazide

  • MDL number MFCD00051848
  • Molecular Weight : 272.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/P10108
Availability details for
Quantity
  • Availability
  • Add to Cart