MAN182107647 Manchester Organics

2,5-Diamino-3-fluoro-4-(trifluoromethyl)pyridine

  • MDL number MFCD04038327
  • Cas Number : 675602-90-1
  • Molecular Weight : 195.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-4-(trifluoromethyl)-2,5-pyridinediamine
Inchi Code 1S/C6H5F4N3/c7-4-3(6(8,9)10)2(11)1-13-5(4)12/h1H,11H2,(H2,12,13)
Inchi Key SMAJZLPBXNXMJE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/S10346
Availability details for
Quantity
  • Availability
  • Add to Cart