MAN182107648 Manchester Organics

2,6-Diamino-3-fluoro-4-(trifluoromethyl)pyridine

  • MDL number MFCD08687525
  • Molecular Weight : 195.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin GB
Product Link https://www.manchesterorganics.com/P10605
Availability details for
Quantity
  • Availability
  • Add to Cart