MAN182107650 Manchester Organics

2,4-Dimethylbenzenesulfonamide

  • MDL number MFCD00460793
  • Cas Number : 7467-12-1
  • Molecular Weight : 185.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/A10876
Availability details for
Quantity
  • Availability
  • Add to Cart