MAN182107654 Manchester Organics

3,5-Dichlorophenylhydrazine

  • MDL number MFCD00051527
  • Cas Number : 39943-56-1
  • Molecular Weight : 177.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/R10228
Availability details for
Quantity
  • Availability
  • Add to Cart