MAN182107660 Manchester Organics

2,2-Difluoro-N-methoxy-N-methylacetamide

  • MDL number MFCD06248152
  • Molecular Weight : 139.101

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V10445
Availability details for
Quantity
  • Availability
  • Add to Cart