MAN182107672 Manchester Organics

3,4-Difluorophenylglyoxal hydrate

  • MDL number MFCD03453081
  • Cas Number : 79784-34-2
  • Molecular Weight : 188.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-difluorophenyl)(oxo)acetaldehyde hydrate
Inchi Code 1S/C8H4F2O2.H2O/c9-6-2-1-5(3-7(6)10)8(12)4-11;/h1-4H;1H2
Inchi Key FFYIMFMDGMXVOF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/N10414
Availability details for
Quantity
  • Availability
  • Add to Cart