MAN182107698 Manchester Organics

5-(4'-Fluorophenyl)-5-oxopentanoic acid

  • MDL number MFCD00667222
  • Cas Number : 149437-76-3
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(4-fluorophenyl)-5-oxopentanoic acid
Inchi Code 1S/C11H11FO3/c12-9-6-4-8(5-7-9)10(13)2-1-3-11(14)15/h4-7H,1-3H2,(H,14,15)
Inchi Key ZBQROUOOMAMCQW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/X10115
Availability details for
Quantity
  • Availability
  • Add to Cart