MAN182107729 Manchester Organics

4-(Trifluoromethyl)maleic anhydride

  • MDL number MFCD00153287
  • Molecular Weight : 166.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F16868

Safety Information

MSDS https://www.manchesterorganics.com/F16868
Availability details for
Quantity
  • Availability
  • Add to Cart