MAN182107734 Manchester Organics

2-Propylbenzene-1,3-diol

  • MDL number MFCD00100568
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z11017

Safety Information

MSDS https://www.manchesterorganics.com/Z11017
Availability details for
Quantity
  • Availability
  • Add to Cart