MAN182107767 Manchester Organics

4-Chloro-2,5-dimethylaniline

  • MDL number MFCD08445705
  • Molecular Weight : 155.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B10570
Availability details for
Quantity
  • Availability
  • Add to Cart