MAN182107771 Manchester Organics

5-Chloro-2-methoxy-4-methylbenzenesulfonamide

  • MDL number MFCD08445708
  • Cas Number : 199590-75-5
  • Molecular Weight : 235.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/H10647
Availability details for
Quantity
  • Availability
  • Add to Cart