MAN182107776 Manchester Organics

Diethyl-4-fluoroheptanedioate

  • MDL number MFCD08704574
  • Molecular Weight : 234.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/B10616
Availability details for
Quantity
  • Availability
  • Add to Cart