MAN182107777 Manchester Organics

2,2-Difluoro-N-(2-hydroxyethyl)propionamide

  • MDL number MFCD08687553
  • Cas Number : 851728-91-1
  • Molecular Weight : 153.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2-difluoro-N-(2-hydroxyethyl)propanamide
Inchi Code 1S/C5H9F2NO2/c1-5(6,7)4(10)8-2-3-9/h9H,2-3H2,1H3,(H,8,10)
Inchi Key QGRLXCZSOWEORV-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/T10585
Availability details for
Quantity
  • Availability
  • Add to Cart