MAN182107779 Manchester Organics

2,4-Dimethoxybenzenesulfonamide

  • MDL number MFCD06147001
  • Cas Number : 51770-71-9
  • Molecular Weight : 217.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,4-dimethoxybenzenesulfonamide
Inchi Code 1S/C8H11NO4S/c1-12-6-3-4-8(14(9,10)11)7(5-6)13-2/h3-5H,1-2H3,(H2,9,10,11)
Inchi Key MGHCDRVTMABICG-UHFFFAOYSA-N

Description

Purity 96%
Country of Origin GB
Product Link https://www.manchesterorganics.com/X10731
Availability details for
Quantity
  • Availability
  • Add to Cart